Assay Details
Target Gene Details
Entrez Gene ID: | 196403 |
Gene Name: | deltex E3 ubiquitin ligase 3 |
Gene Aliases: |
RNF154, deltex3 |
Location: |
Chr.12:57604327-57609804 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTX3 | NM_001286245.1 | NP_001273174.1 | ||
| NM_001286246.1 | NP_001273175.1 | |||
| NM_178502.3 | 2 | 293 | NP_848597.1 | |
| XM_005268697.1 | 2 | 815 | XP_005268754.1 | |
| XM_005268698.1 | 2 | 588 | XP_005268755.1 | |
| XM_005268700.1 | XP_005268757.1 | |||
| XM_005268703.1 | XP_005268760.1 | |||
| XM_011538022.2 | 1 | 671 | XP_011536324.1 | |
| AK092085.1 | 2 | 94 | BAC03801.1 | |
| AK094385.1 | BAC04344.1 | |||
| AK128752.1 | 1 | 803 | ||
| AL831941.1 | 1 | 620 | CAD38593.2 | |
| BC114441.1 | ||||
| BC114498.1 | ||||
| DA684401.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain |
|
| nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss |
|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map