Assay Details
Target Gene Details
Entrez Gene ID: | 220965 |
Gene Name: | family with sequence similarity 13 member C |
Gene Aliases: |
FAM13C1 |
Location: |
Chr.10:59245933-59363186 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831888 | Chr.10:59183895 - 59378493 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map