Assay Details
Target Gene Details
Entrez Gene ID: | 348110 |
Gene Name: | actin-related protein 2/3 complex inhibitor |
Gene Aliases: |
C15orf38 |
Location: |
Chr.15:89896532-89912990 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPIN | NM_001282380.1 | 6 | 1378 | NP_001269309.1 |
| NM_182616.3 | 6 | 989 | NP_872422.1 | |
| AK093274.1 | 6 | 1378 | ||
| BC053602.1 | 6 | 926 | AAH53602.1 |
Target Gene Details
Entrez Gene ID: | 100526783 |
Gene Name: | C15orf38-AP3S2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.15:89830599-89912990 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C15orf38-AP3S2 | NM_001199058.1 | NP_001185987.1 | ||
| AK000495.1 |
More Information
Set Membership: |
|

Genomic Map