Assay Details
Target Gene Details
Entrez Gene ID: | 29083 |
Gene Name: | GTP binding protein 8 (putative) |
Gene Aliases: |
HSPC135 |
Location: |
Chr.3:112989275-113001966 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP8 | NM_014170.2 | 6 | 1042 | NP_054889.2 |
| NM_138485.1 | 5 | 943 | NP_612494.1 | |
| AI367201.1 | ||||
| AK289611.1 | 5 | 905 | ||
| AK292651.1 | 6 | 1061 | ||
| BX354218.2 | ||||
| CA429378.1 | 1 | 165 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591293 | Chr.3:112266260 - 113293950 on Build GRCh38 | Loss |
|
| esv3893763 | Chr.3:112998472 - 113113216 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map