Assay Details
Target Gene Details
Entrez Gene ID: | 10011 |
Gene Name: | steroid receptor RNA activator 1 |
Gene Aliases: |
SRA, SRAP, STRAA1, pp7684 |
Location: |
Chr.5:140550067-140558093 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRA1 | NM_001035235.3 | 5 | 1509 | NP_001030312.2 |
| NM_001253764.1 | 3 | 824 | NP_001240693.1 | |
| NR_045586.1 | 5 | 1569 | ||
| NR_045587.1 | 4 | 1027 | ||
| AF092038.1 | ||||
| AF318361.1 | AAL55868.1 | |||
| AI140060.1 | ||||
| AK024640.1 | 4 | 1013 | ||
| AK054960.1 | 5 | 1729 | ||
| BC040043.2 | AAH40043.2 | |||
| BQ052761.1 | 1 | 261 | ||
| BU553214.1 |
More Information
Set Membership: |
|

Genomic Map