Assay Details
Target Gene Details
Entrez Gene ID: | 4158 |
Gene Name: | melanocortin 2 receptor |
Gene Aliases: |
ACTHR |
Location: |
Chr.18:13882042-13915707 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MC2R | NM_000529.2 | 2 | 3426 | NP_000520.1 |
| NM_001291911.1 | 2 | 3455 | NP_001278840.1 | |
| XM_017025781.1 | 3 | 4048 | XP_016881270.1 | |
| Y10259.1 | 1 | 2358 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066142 | Chr.18:13801920 - 14221947 on Build GRCh38 | Loss |
|
| nsv576529 | Chr.18:13606185 - 14150725 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map