Assay Details
Target Gene Details
Entrez Gene ID: | 3843 |
Gene Name: | importin 5 |
Gene Aliases: |
IMB3, KPNB3, Pse1, RANBP5, imp5 |
Location: |
Chr.13:97953641-98024296 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPO5 | NM_002271.4 | 9 | 849 | NP_002262.3 |
| XM_005254052.3 | 8 | 759 | XP_005254109.1 | |
| XM_005254053.3 | 8 | 776 | XP_005254110.1 | |
| XM_011521087.2 | 10 | 940 | XP_011519389.1 | |
| XM_011521088.2 | 10 | 1006 | XP_011519390.1 | |
| XM_011521089.2 | 11 | 1063 | XP_011519391.1 | |
| XM_011521090.2 | 9 | 899 | XP_011519392.1 | |
| XM_017020562.1 | 9 | 880 | XP_016876051.1 | |
| AK225286.1 | 6 | 634 | ||
| AK302812.1 | 6 | 621 | ||
| AK303494.1 | ||||
| BC001497.2 | 6 | 617 | AAH01497.1 | |
| BC019309.1 | 6 | 617 | AAH19309.1 | |
| BC045640.1 | 9 | 830 | AAH45640.1 | |
| BX648222.1 | 6 | 679 | ||
| U72761.1 | 6 | 615 | AAC51317.1 | |
| Y08890.1 | 9 | 851 | CAA70103.1 |
More Information
Set Membership: |
|

Genomic Map