Assay Details
Target Gene Details
Entrez Gene ID: | 54 |
Gene Name: | acid phosphatase 5, tartrate resistant |
Gene Aliases: |
HPAP, TRACP5a, TRACP5b, TRAP, TrATPase |
Location: |
Chr.19:11574660-11579008 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACP5 | NM_001111034.2 | NP_001104504.1 | ||
| NM_001111035.2 | NP_001104505.1 | |||
| NM_001111036.2 | NP_001104506.1 | |||
| NM_001322023.1 | NP_001308952.1 | |||
| NM_001611.4 | NP_001602.1 | |||
| XM_005259938.1 | XP_005259995.1 | |||
| XM_011528069.2 | XP_011526371.1 | |||
| AK225912.1 | ||||
| AK290717.1 | ||||
| BC025414.2 | AAH25414.1 | |||
| BC111014.1 | AAI11015.1 | |||
| BI755033.1 | ||||
| BX365007.2 | ||||
| CR457078.1 | CAG33359.1 | |||
| J04430.1 | AAA76849.1 | |||
| X14618.1 | CAA32771.1 |
Target Gene Details
Entrez Gene ID: | 199692 |
Gene Name: | zinc finger protein 627 |
Gene Aliases: |
- |
Location: |
Chr.19:11575255-11619159 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF627 | XM_011527780.2 | XP_011526082.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509721 | Chr.19:11525216 - 11691390 on Build GRCh38 | Insertion |
|
| nsv519802 | Chr.19:11236817 - 11578343 on Build GRCh38 | Loss |
|
| esv991647 | Chr.19:11575212 - 11587674 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map