Assay Details
Target Gene Details
Entrez Gene ID: | 55357 |
Gene Name: | TBC1 domain family member 2 |
Gene Aliases: |
PARIS-1, PARIS1, TBC1D2A |
Location: |
Chr.9:98198998-98255721 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D2 | NM_001267571.1 | NP_001254500.1 | ||
| NM_001267572.1 | NP_001254501.1 | |||
| NM_018421.3 | NP_060891.3 | |||
| NR_051978.1 | ||||
| XM_011518843.1 | XP_011517145.1 | |||
| XM_017014899.1 | XP_016870388.1 | |||
| AB449882.1 | ||||
| AF318370.1 | AAL55877.1 | |||
| AK026105.1 | BAB15361.1 | |||
| AK124772.1 | ||||
| AK131290.1 | ||||
| AK298575.1 | ||||
| AL353935.1 | CAB89247.2 | |||
| AY026527.1 | AAK07684.1 | |||
| BC028918.1 | AAH28918.1 | |||
| BC071978.1 | AAH71978.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520067 | Chr.9:98164879 - 98216260 on Build GRCh38 | Gain |
|
| esv3621201 | Chr.9:98068567 - 98407578 on Build GRCh38 | Gain |
|
| nsv831663 | Chr.9:98178766 - 98347210 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map