Assay Details
Target Gene Details
Entrez Gene ID: | 125113 |
Gene Name: | keratin 222 |
Gene Aliases: |
KA21, KRT222P |
Location: |
Chr.17:40655620-40665164 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT222 | NM_152349.2 | 3 | 329 | NP_689562.1 |
| AK308598.1 | 3 | 393 | ||
| BC032815.1 | 3 | 288 | AAH32815.1 | |
| BX538083.1 | 3 | 282 | CAD98007.1 | |
| DB465511.1 | 3 | 329 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574994 | Chr.17:40563547 - 40668384 on Build GRCh38 | Gain |
|
| esv2758689 | Chr.17:40592048 - 40693852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map