Assay Details
Target Gene Details
Entrez Gene ID: | 80321 |
Gene Name: | centrosomal protein 70 |
Gene Aliases: |
BITE |
Location: |
Chr.3:138494339-138594383 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP70 | NM_001288964.1 | 18 | 2513 | NP_001275893.1 |
| NM_001288965.1 | 17 | 2457 | NP_001275894.1 | |
| NM_001288967.1 | 14 | 2061 | NP_001275896.1 | |
| NM_001320598.1 | 18 | 2583 | NP_001307527.1 | |
| NM_024491.3 | 18 | 2531 | NP_077817.2 | |
| XM_017007275.1 | 18 | 2867 | XP_016862764.1 | |
| XM_017007282.1 | 17 | 2332 | XP_016862771.1 | |
| XM_017007283.1 | 17 | 2263 | XP_016862772.1 | |
| XM_017007284.1 | 11 | 2567 | XP_016862773.1 | |
| AF202146.1 | 18 | 2385 | AAG35616.1 | |
| AK023098.1 | 16 | 2246 | BAB14403.1 | |
| AK299003.1 | 17 | 2322 | ||
| BC030598.2 | 18 | 2433 | AAH30598.1 | |
| BF435951.1 | 1 | 247 | ||
| BU634409.1 | 1 | 244 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999041 | Chr.3:138429970 - 138629146 on Build GRCh38 | Gain |
|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
| esv3597916 | Chr.3:138478438 - 138638584 on Build GRCh38 | Gain |
|
| dgv4908n100 | Chr.3:138494324 - 139018846 on Build GRCh38 | Gain |
|
| nsv522517 | Chr.3:138455681 - 138579140 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map