Assay Details
Target Gene Details
Entrez Gene ID: | 78988 |
Gene Name: | mitochondrial ribosomal protein L57 |
Gene Aliases: |
MRP63, bMRP63 |
Location: |
Chr.13:21176233-21179084 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL57 | NM_024026.4 | 1 | 362 | NP_076931.1 |
| AW445203.1 | 1 | 93 |
Target Gene Details
Entrez Gene ID: | 221150 |
Gene Name: | spindle and kinetochore associated complex subunit 3 |
Gene Aliases: |
C13orf3, RAMA1 |
Location: |
Chr.13:21153595-21176602 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKA3 | NM_001166017.1 | NP_001159489.1 | ||
| NM_145061.5 | NP_659498.4 | |||
| XM_011534994.1 | XP_011533296.1 | |||
| AF361358.1 | AAO13796.1 | |||
| AK090584.1 | BAC03484.1 | |||
| AK307976.1 | ||||
| AK314864.1 | ||||
| BC048988.1 | AAH48988.1 | |||
| BI460315.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037082 | Chr.13:20932196 - 21204957 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map