Assay Details
Target Gene Details
Entrez Gene ID: | 25957 |
Gene Name: | PNN interacting serine and arginine rich protein |
Gene Aliases: |
C6orf111, HSPC306, SFRS18, SRrp130, bA98I9.2 |
Location: |
Chr.6:99398051-99425345 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890914 | Chr.6:99393043 - 99409897 on Build GRCh38 | Loss |
|
| nsv527517 | Chr.6:99394180 - 99416102 on Build GRCh38 | Loss |
|
| nsv1029536 | Chr.6:98422726 - 99818247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map