Assay Details
Target Gene Details
Entrez Gene ID: | 56300 |
Gene Name: | interleukin 36, gamma |
Gene Aliases: |
IL-1F9, IL-1H1, IL-1RP2, IL1E, IL1F9, IL1H1, IL1RP2 |
Location: |
Chr.2:112978019-112985672 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL36G | NM_001278568.1 | NP_001265497.1 | ||
| NM_019618.3 | NP_062564.1 | |||
| AF200492.1 | AAF69248.1 | |||
| AF206696.1 | AAG35670.1 | |||
| BC096721.1 | AAH96721.1 | |||
| BC098130.1 | AAH98130.1 | |||
| BC098155.1 | AAH98155.1 | |||
| BC098337.1 | AAH98337.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2880 | Chr.2:112959225 - 112979344 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map