Assay Details
Target Gene Details
Entrez Gene ID: | 9564 |
Gene Name: | BCAR1, Cas family scaffolding protein |
Gene Aliases: |
CAS, CAS1, CASS1, CRKAS, P130Cas |
Location: |
Chr.16:75229030-75268053 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv516504 | Chr.16:75189593 - 75272504 on Build GRCh38 | Gain+Loss |
|
| nsv833282 | Chr.16:75120273 - 75296736 on Build GRCh38 | Gain+Loss |
|
| esv3582321 | Chr.16:75232709 - 75427727 on Build GRCh38 | Loss |
|
| nsv523405 | Chr.16:75194071 - 75308948 on Build GRCh38 | Loss |
|
| nsv471095 | Chr.16:75235635 - 75286929 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map