Assay Details
Target Gene Details
Entrez Gene ID: | 51111 |
Gene Name: | lysine methyltransferase 5B |
Gene Aliases: |
CGI-85, CGI85, SUV420H1 |
Location: |
Chr.11:68154863-68213904 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KMT5B | NM_001300907.1 | 12 | 2330 | NP_001287836.1 |
| NM_001300908.1 | 11 | 2297 | NP_001287837.1 | |
| NM_017635.4 | 11 | 2262 | NP_060105.3 | |
| XM_005274035.3 | 11 | 2482 | XP_005274092.2 | |
| XM_005274036.3 | 10 | 2413 | XP_005274093.2 | |
| XM_006718581.1 | 10 | 2144 | XP_006718644.1 | |
| XM_011545091.1 | 10 | 2056 | XP_011543393.1 | |
| XM_011545092.2 | 10 | 2246 | XP_011543394.1 | |
| XM_011545093.2 | 11 | 2527 | XP_011543395.1 | |
| XM_017017876.1 | 12 | 2843 | XP_016873365.1 | |
| XM_017017877.1 | 12 | 2629 | XP_016873366.1 | |
| XM_017017878.1 | 12 | 2574 | XP_016873367.1 | |
| XM_017017879.1 | 12 | 2375 | XP_016873368.1 | |
| AK000046.1 | 1 | 657 | BAA90905.1 | |
| AK025461.1 | 6 | 1386 | ||
| AK026428.1 | 6 | 1372 | ||
| AK225203.1 | 1 | 657 | ||
| AK314336.1 | 11 | 2124 | ||
| AL512763.1 | 1 | 207 | CAC21680.1 | |
| BC012933.1 | 3 | 1017 | ||
| BC098121.1 | 10 | 2028 | AAH98121.1 | |
| BC099714.1 | 10 | 1983 | AAH99714.1 | |
| BC103498.1 | 10 | 1983 | AAI03499.1 | |
| BC104483.1 | 10 | 1983 | AAI04484.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825963 | Chr.11:67996205 - 68429586 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map