Assay Details
Target Gene Details
Entrez Gene ID: | 27440 |
Gene Name: | cat eye syndrome chromosome region, candidate 5 |
Gene Aliases: |
- |
Location: |
Chr.22:17137520-17165287 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CECR5 | NM_017829.5 | NP_060299.4 | ||
| AF273270.1 | AAK19151.1 |
Target Gene Details
Entrez Gene ID: | 100130717 |
Gene Name: | CECR5 antisense RNA 1 |
Gene Aliases: |
CECR4, NCRNA00017 |
Location: |
Chr.22:17159389-17165445 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CECR5-AS1 | NR_024482.1 | |||
| NR_024483.1 | ||||
| AF307448.1 | AAK30048.1 | |||
| AW293178.1 | 1 | 161 | ||
| BI962745.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828924 | Chr.22:17116393 - 17174153 on Build GRCh38 | Gain |
|
| esv3647209 | Chr.22:17151829 - 17264992 on Build GRCh38 | Gain |
|
| nsv1064641 | Chr.22:16757380 - 17233805 on Build GRCh38 | Gain |
|
| nsv509805 | Chr.22:17081417 - 17267685 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map