Assay Details
Target Gene Details
Entrez Gene ID: | 121391 |
Gene Name: | keratin 74 |
Gene Aliases: |
ADWH, HTSS2, HYPT3, K6IRS4, KRT5C, KRT6IRS4 |
Location: |
Chr.12:52565819-52573825 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT74 | NM_175053.3 | 6 | 1127 | NP_778223.2 |
| AJ508777.1 | 6 | 1127 | CAD48514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832415 | Chr.12:52567563 - 52757300 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map