Assay Details
Target Gene Details
Entrez Gene ID: | 79760 |
Gene Name: | gem nuclear organelle associated protein 7 |
Gene Aliases: |
SIP3 |
Location: |
Chr.19:45075728-45091524 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GEMIN7 | NM_001007269.1 | 2 | 1318 | NP_001007270.1 |
| NM_001007270.1 | 3 | 1355 | NP_001007271.1 | |
| NM_024707.2 | 3 | 1441 | NP_078983.1 | |
| AK024018.1 | 3 | 1425 | BAB14780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828563 | Chr.19:45091333 - 45094910 on Build GRCh38 | Gain |
|
| nsv953587 | Chr.19:45039143 - 45166142 on Build GRCh38 | Deletion |
|
| esv3644497 | Chr.19:45085975 - 45092273 on Build GRCh38 | Loss |
|
| nsv833843 | Chr.19:45090630 - 45290424 on Build GRCh38 | Loss |
|
| esv27359 | Chr.19:45085653 - 45092119 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map