Assay Details
Target Gene Details
Entrez Gene ID: | 80167 |
Gene Name: | abhydrolase domain containing 18 |
Gene Aliases: |
C4orf29 |
Location: |
Chr.4:127965256-128039711 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD18 | NM_001319305.1 | 1 | 208 | NP_001306234.1 |
| NM_001319306.1 | 1 | 208 | NP_001306235.1 | |
| NM_001319307.1 | 1 | 208 | NP_001306236.1 | |
| XM_005263238.4 | 1 | 203 | XP_005263295.1 | |
| XM_006714318.3 | 1 | 203 | XP_006714381.1 | |
| XM_006714320.3 | XP_006714383.1 | |||
| XM_011532272.2 | 1 | 167 | XP_011530574.1 | |
| XM_011532273.2 | 1 | 167 | XP_011530575.1 | |
| XM_011532274.2 | 1 | 203 | XP_011530576.1 | |
| XM_011532276.1 | 1 | 203 | XP_011530578.1 | |
| XM_011532278.2 | 1 | 167 | XP_011530580.1 | |
| XM_017008631.1 | 1 | 203 | XP_016864120.1 | |
| AK024759.1 | 1 | 58 | BAB14990.1 | |
| BC034253.1 | 1 | 87 | ||
| BX500590.1 | 1 | 181 | ||
| HY086029.1 | 1 | 209 |
Target Gene Details
Entrez Gene ID: | 256471 |
Gene Name: | major facilitator superfamily domain containing 8 |
Gene Aliases: |
CCMD, CLN7 |
Location: |
Chr.4:127917805-127965984 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD8 | NM_152778.2 | NP_689991.1 | ||
| AK294184.1 | ||||
| AK294527.1 | ||||
| AK294957.1 | ||||
| BC029503.1 | AAH29503.1 | |||
| BI553701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5385n100 | Chr.4:127882004 - 128173863 on Build GRCh38 | Gain |
|
| nsv595388 | Chr.4:127932758 - 128163676 on Build GRCh38 | Gain |
|
| nsv1019193 | Chr.4:127889372 - 128028165 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map