Assay Details
Target Gene Details
Entrez Gene ID: | 3751 |
Gene Name: | potassium voltage-gated channel subfamily D member 2 |
Gene Aliases: |
KV4.2, RK5 |
Location: |
Chr.7:120273635-120750333 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCND2 | NM_012281.2 | 1 | 726 | NP_036413.1 |
| XM_011516165.2 | 1 | 759 | XP_011514467.1 | |
| AB028967.1 | 1 | 726 | ||
| AJ010969.1 | 1 | 190 | CAB56841.1 | |
| AK307139.1 | 1 | 759 | ||
| BC110449.1 | 1 | 380 | AAI10450.1 | |
| BC110450.1 | 1 | 380 | AAI10451.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608326 | Chr.7:120188277 - 120355836 on Build GRCh38 | Loss |
|
| nsv1031971 | Chr.7:119385956 - 120677299 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map