Assay Details
Target Gene Details
Entrez Gene ID: | 51277 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C27 |
Gene Aliases: |
RBJ, RabJS |
Location: |
Chr.2:24943636-24972094 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC27 | NM_001198559.1 | 6 | 3706 | NP_001185488.1 |
| NM_016544.2 | 7 | 3867 | NP_057628.1 | |
| XM_017004269.1 | 7 | 3898 | XP_016859758.1 | |
| XM_017004270.1 | 7 | 3758 | XP_016859759.1 | |
| AL117490.1 | 1 | 2689 | ||
| BC034049.1 | 7 | 3728 | AAH34049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004529 | Chr.2:24363242 - 25005468 on Build GRCh38 | Loss |
|
| nsv1008732 | Chr.2:24907673 - 25349661 on Build GRCh38 | Loss |
|
| nsv997725 | Chr.2:24052436 - 25123406 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map