Assay Details
Target Gene Details
Entrez Gene ID: | 369 |
Gene Name: | A-Raf proto-oncogene, serine/threonine kinase |
Gene Aliases: |
A-RAF, ARAF1, PKS2, RAFA1 |
Location: |
Chr.X:47561100-47571921 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARAF | NM_001256196.1 | NP_001243125.1 | ||
| NM_001654.4 | NP_001645.1 | |||
| XM_006724529.2 | XP_006724592.1 | |||
| XM_011543906.2 | XP_011542208.1 | |||
| XM_011543908.2 | XP_011542210.1 | |||
| XM_011543909.2 | XP_011542211.1 | |||
| AB208831.1 | BAD92068.1 | |||
| AF052136.1 | ||||
| AK130043.1 | ||||
| BC002466.2 | AAH02466.1 | |||
| BC007514.2 | AAH07514.1 | |||
| X04790.1 | CAA28476.1 |
Target Gene Details
Entrez Gene ID: | 6853 |
Gene Name: | synapsin I |
Gene Aliases: |
SYN1a, SYN1b, SYNI |
Location: |
Chr.X:47571901-47619857 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYN1 | NM_006950.3 | NP_008881.2 | ||
| NM_133499.2 | NP_598006.1 | |||
| AL833961.1 |
More Information
Set Membership: |
|

Genomic Map