Assay Details
Target Gene Details
Entrez Gene ID: | 7089 |
Gene Name: | transducin like enhancer of split 2 |
Gene Aliases: |
ESG, ESG2, GRG2 |
Location: |
Chr.19:2997638-3047635 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLE2 | NM_001144761.1 | 20 | 2209 | NP_001138233.1 |
| NM_001144762.1 | 18 | 2000 | NP_001138234.1 | |
| NM_001300846.1 | 20 | 2460 | NP_001287775.1 | |
| NM_003260.4 | 20 | 2423 | NP_003251.2 | |
| XM_006722864.2 | 19 | 2291 | XP_006722927.1 | |
| XM_011528230.1 | 20 | 2332 | XP_011526532.1 | |
| AK123183.1 | 9 | 1714 | ||
| AK293407.1 | 18 | 2002 | ||
| AK293482.1 | 16 | 2252 | ||
| AK308137.1 | 20 | 2208 | ||
| AK312285.1 | 20 | 2272 | ||
| BC017364.2 | 20 | 2280 | AAH17364.1 | |
| M99436.1 | 20 | 2185 | AAA61193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833709 | Chr.19:2950244 - 3068609 on Build GRCh38 | Loss |
|
| dgv47n68 | Chr.19:2977776 - 3162121 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map