Assay Details
Target Gene Details
Entrez Gene ID: | 332 |
Gene Name: | baculoviral IAP repeat containing 5 |
Gene Aliases: |
API4, EPR-1 |
Location: |
Chr.17:78214196-78225635 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BIRC5 | NM_001012270.1 | 3 | 850 | NP_001012270.1 |
| NM_001012271.1 | 5 | 1037 | NP_001012271.1 | |
| NM_001168.2 | 4 | 968 | NP_001159.2 | |
| AF077350.1 | 4 | 871 | AAD34226.1 | |
| AK223428.1 | 4 | 913 | BAD97148.1 | |
| AK301793.1 | 5 | 1086 | ||
| AL547142.3 | 3 | 782 | ||
| BC008718.2 | 4 | 880 | AAH08718.1 | |
| BC034148.1 | 4 | 891 | AAH34148.1 | |
| BC065497.1 | 4 | 861 | AAH65497.1 | |
| BG178974.1 | 3 | 766 | ||
| BG759111.1 | 1 | 519 | ||
| BQ687404.1 | 4 | 745 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524306 | Chr.17:78183449 - 78280693 on Build GRCh38 | Gain |
|
| nsv952373 | Chr.17:78214820 - 78283319 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map