Assay Details
Target Gene Details
Entrez Gene ID: | 90203 |
Gene Name: | sorting nexin family member 21 |
Gene Aliases: |
C20orf161, PP3993, SNX-L, dJ337O18.4 |
Location: |
Chr.20:45833418-45843275 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNX21 | NM_001042632.2 | 4 | 1342 | NP_001036097.1 |
| NM_001042633.2 | 5 | 1360 | NP_001036098.1 | |
| NM_033421.3 | 4 | 1349 | NP_219489.1 | |
| NM_152897.2 | NP_690857.1 | |||
| XM_005260608.4 | 2 | 1424 | XP_005260665.1 | |
| XM_011529095.2 | 2 | 1328 | XP_011527397.1 | |
| AF258570.1 | 1 | 1009 | AAG23773.1 | |
| AF523834.1 | 4 | 1328 | AAM77915.1 | |
| AK091359.1 | 3 | 1364 | BAC03646.1 | |
| AK095851.1 | 3 | 1223 | BAC04637.1 | |
| AK226120.1 | ||||
| AK293443.1 | ||||
| AL591562.1 | 4 | 1328 | CAC39140.1 | |
| BC012904.1 | 6 | 1580 | ||
| BC019823.1 | AAH19823.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833990 | Chr.20:45771510 - 45946844 on Build GRCh38 | Loss |
|
| nsv586132 | Chr.20:45785138 - 45842587 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map