Assay Details
Target Gene Details
Entrez Gene ID: | 84836 |
Gene Name: | abhydrolase domain containing 14B |
Gene Aliases: |
CIB, HEL-S-299 |
Location: |
Chr.3:51968510-51974630 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD14B | NM_001146314.1 | NP_001139786.1 | ||
| NM_001254753.1 | NP_001241682.1 | |||
| NM_032750.2 | 2 | 867 | NP_116139.1 | |
| AK075112.1 | 2 | 258 | BAC11408.1 | |
| AK096073.1 | BAC04696.1 | |||
| AK296599.1 | ||||
| AK298022.1 | ||||
| AK298813.1 | ||||
| BC007234.1 | 2 | 251 | AAH07234.1 | |
| BC050650.1 | AAH50650.1 | |||
| BC056411.1 | AAH56411.1 | |||
| DA315486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003453 | Chr.3:51928065 - 52065425 on Build GRCh38 | Gain |
|
| nsv508922 | Chr.3:51940932 - 52009520 on Build GRCh38 | Insertion |
|
| nsv954857 | Chr.3:51932485 - 52056584 on Build GRCh38 | Deletion |
|
| nsv519445 | Chr.3:51970301 - 52333916 on Build GRCh38 | Gain+Loss |
|
| nsv520211 | Chr.3:51738331 - 52132131 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78583983] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map