Assay Details
Target Gene Details
Entrez Gene ID: | 9813 |
Gene Name: | EF-hand calcium binding domain 14 |
Gene Aliases: |
KIAA0494 |
Location: |
Chr.1:46675159-46719064 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFCAB14 | NM_014774.2 | NP_055589.1 | ||
| AB007963.1 | ||||
| AK295722.1 | ||||
| BC002525.2 | AAH02525.1 |
More Information
Set Membership: |
|

Genomic Map