Assay Details
Target Gene Details
Entrez Gene ID: | 440757 |
Gene Name: | long intergenic non-protein coding RNA 851 |
Gene Aliases: |
DZANK1-AS1 |
Location: |
Chr.20:18379049-18381484 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00851 | NR_034167.1 | 2 | 633 | |
| BC033985.1 | 2 | 638 | AAH33985.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509763 | Chr.20:18299024 - 18383464 on Build GRCh38 | Insertion |
|
| esv3645437 | Chr.20:18357770 - 18484726 on Build GRCh38 | Gain |
|
| dgv4283n100 | Chr.20:18364565 - 18647377 on Build GRCh38 | Loss |
|
| nsv1062244 | Chr.20:18184227 - 18520274 on Build GRCh38 | Gain |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map