Assay Details
Target Gene Details
Entrez Gene ID: | 2346 |
Gene Name: | folate hydrolase (prostate-specific membrane antigen) 1 |
Gene Aliases: |
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP |
Location: |
Chr.11:49145094-49208706 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOLH1 | NM_001014986.1 | 1 | 54 | NP_001014986.1 |
| NM_001193471.1 | 1 | 54 | NP_001180400.1 | |
| NM_001193472.1 | 1 | 54 | NP_001180401.1 | |
| NM_001193473.1 | 1 | 54 | NP_001180402.1 | |
| NM_004476.1 | 1 | 54 | NP_004467.1 | |
| XM_011519958.2 | XP_011518260.2 | |||
| XM_017017432.1 | XP_016872921.1 | |||
| XM_017017433.1 | XP_016872922.1 | |||
| XM_017017434.1 | XP_016872923.1 | |||
| XM_017017435.1 | XP_016872924.1 | |||
| XM_017017436.1 | XP_016872925.1 | |||
| XM_017017438.1 | XP_016872927.1 | |||
| XM_017017439.1 | XP_016872928.1 | |||
| XM_017017440.1 | XP_016872929.1 | |||
| XM_017017442.1 | 1 | 90 | XP_016872931.1 | |
| XM_017017444.1 | XP_016872933.1 | |||
| XM_017017445.1 | XP_016872934.1 | |||
| XM_017017446.1 | XP_016872935.1 | |||
| XM_017017447.1 | XP_016872936.1 | |||
| XM_017017449.1 | XP_016872938.1 | |||
| XM_017017450.1 | XP_016872939.1 | |||
| XM_017017451.1 | XP_016872940.1 | |||
| AK295368.1 | ||||
| AK295470.1 | ||||
| AK300532.1 | ||||
| AK312366.1 | ||||
| M99487.1 | 1 | 54 | AAA60209.1 | |
| S76978.1 | 1 | 54 | AAB33750.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv467874 | Chr.11:48483044 - 49935211 on Build GRCh38 | Gain |
|
| nsv554347 | Chr.11:49089915 - 49935211 on Build GRCh38 | Gain |
|
| nsv518571 | Chr.11:49141396 - 49606281 on Build GRCh38 | Gain |
|
| esv2759825 | Chr.11:49190685 - 49447414 on Build GRCh38 | Gain+Loss |
|
| esv3891996 | Chr.11:49173538 - 49443281 on Build GRCh38 | Gain |
|
| nsv1044070 | Chr.11:49126191 - 49427222 on Build GRCh38 | Loss |
|
| esv3891995 | Chr.11:49134487 - 49221694 on Build GRCh38 | Gain |
|
| esv2744485 | Chr.11:48902336 - 49894003 on Build GRCh38 | Deletion |
|
| nsv972015 | Chr.11:49165970 - 49219167 on Build GRCh38 | Duplication |
|
| nsv469956 | Chr.11:48901093 - 49846333 on Build GRCh38 | Gain |
|
| esv3891986 | Chr.11:48021501 - 49282806 on Build GRCh38 | Gain |
|
| esv3891984 | Chr.11:48711661 - 49369067 on Build GRCh38 | Gain |
|
| esv2666171 | Chr.11:48963976 - 49887121 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map