Assay Details
Target Gene Details
Entrez Gene ID: | 5096 |
Gene Name: | propionyl-CoA carboxylase beta subunit |
Gene Aliases: |
- |
Location: |
Chr.3:136250325-136330171 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCCB | NM_000532.4 | 8 | 871 | NP_000523.2 |
| NM_001178014.1 | 9 | 931 | NP_001171485.1 | |
| XM_011512873.1 | 8 | 871 | XP_011511175.1 | |
| AB209009.1 | 8 | 838 | BAD92246.1 | |
| AF217984.1 | 5 | 777 | AAG17227.1 | |
| AK130359.1 | 8 | 829 | ||
| AK225215.1 | 8 | 826 | ||
| AK225733.1 | 8 | 827 | ||
| AK295312.1 | 9 | 898 | ||
| AK302522.1 | 8 | 735 | ||
| AK303079.1 | 6 | 551 | ||
| AL831978.2 | 8 | 821 | CAD89898.1 | |
| BC013768.1 | 8 | 843 | AAH13768.1 | |
| BC053661.1 | 8 | 838 | AAH53661.1 | |
| M13573.1 | AAA60036.1 | |||
| S67325.1 | 8 | 850 | AAB28900.1 | |
| X73424.1 | 8 | 890 | CAA51825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508246 | Chr.3:136203181 - 136319780 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map