Assay Details
Target Gene Details
Entrez Gene ID: | 101928069 |
Gene Name: | uncharacterized LOC101928069 |
Gene Aliases: |
- |
Location: |
Chr.11:66473490-66480249 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928069 | NR_120586.1 | |||
| BQ223044.1 |
Target Gene Details
Entrez Gene ID: | 246330 |
Gene Name: | pellino E3 ubiquitin protein ligase family member 3 |
Gene Aliases: |
- |
Location: |
Chr.11:66466327-66477337 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PELI3 | NM_001098510.1 | 7 | 1933 | NP_001091980.1 |
| NM_001243135.1 | 7 | 1772 | NP_001230064.1 | |
| NM_001243136.1 | 6 | 1780 | NP_001230065.1 | |
| NM_145065.2 | 8 | 2005 | NP_659502.2 | |
| XM_011544884.2 | 8 | 2008 | XP_011543186.1 | |
| XM_017017465.1 | 6 | 1757 | XP_016872954.1 | |
| AF487456.1 | AAO49465.1 | |||
| AF487457.1 | AAO49466.1 | |||
| AK074201.1 | 8 | 1943 | BAB85015.1 | |
| AK075253.1 | 4 | 1445 | BAC11499.1 | |
| AK094060.1 | 7 | 1772 | BAC04275.1 | |
| AL834395.1 | 8 | 1952 | CAD39057.1 | |
| BC025723.1 | 7 | 1854 | AAH25723.1 | |
| BC127759.1 | 2 | 1091 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
| nsv832193 | Chr.11:66352868 - 66511334 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map