Assay Details
Target Gene Details
Entrez Gene ID: | 140838 |
Gene Name: | N-acetylneuraminic acid phosphatase |
Gene Aliases: |
C20orf147, HDHD4, dJ694B14.3 |
Location: |
Chr.20:25612935-25624012 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NANP | NM_152667.2 | 2 | 2078 | NP_689880.1 |
| AK055472.1 | 2 | 2045 | ||
| AL833116.1 | 2 | 2052 | ||
| BC022552.1 | 2 | 2044 | AAH22552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762069 | Chr.20:25525946 - 25638892 on Build GRCh38 | Loss |
|
| nsv3342 | Chr.20:25613217 - 25646797 on Build GRCh38 | Insertion |
|
| esv3645589 | Chr.20:25613861 - 25616080 on Build GRCh38 | Loss |
|
| esv3893324 | Chr.20:25437720 - 25633741 on Build GRCh38 | Gain |
|
| nsv522873 | Chr.20:23923336 - 26134874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map