Assay Details
Target Gene Details
Entrez Gene ID: | 8973 |
Gene Name: | cholinergic receptor nicotinic alpha 6 subunit |
Gene Aliases: |
CHNRA6 |
Location: |
Chr.8:42752620-42768786 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA6 | NM_001199279.1 | NP_001186208.1 | ||
| NM_004198.3 | NP_004189.1 | |||
| AK298798.1 | ||||
| AK313521.1 | ||||
| BC014456.1 | AAH14456.1 | |||
| U62435.1 | AAB40113.1 | |||
| Y16282.1 | CAA76155.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483102 | Chr.8:42722031 - 42876093 on Build GRCh38 | Loss |
|
| dgv7186n100 | Chr.8:42688183 - 42763570 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map