Assay Details
Target Gene Details
Entrez Gene ID: | 5794 |
Gene Name: | protein tyrosine phosphatase, receptor type H |
Gene Aliases: |
R-PTP-H, SAP1 |
Location: |
Chr.19:55181247-55209506 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRH | NM_001161440.2 | 3 | 279 | NP_001154912.2 |
| NM_002842.4 | 3 | 279 | NP_002833.4 | |
| XM_011527183.1 | 3 | 300 | XP_011525485.1 | |
| XM_011527188.1 | 2 | 249 | XP_011525490.1 | |
| XM_011527190.1 | 3 | 300 | XP_011525492.1 | |
| XM_017027056.1 | 2 | 287 | XP_016882545.1 | |
| XM_017027057.1 | 2 | 287 | XP_016882546.1 | |
| XM_017027058.1 | 2 | 320 | XP_016882547.1 | |
| XM_017027059.1 | 2 | 287 | XP_016882548.1 | |
| XM_017027060.1 | 2 | 287 | XP_016882549.1 | |
| XM_017027061.1 | 2 | 287 | XP_016882550.1 | |
| XM_017027062.1 | 2 | 195 | XP_016882551.1 | |
| XM_017027063.1 | 2 | 188 | XP_016882552.1 | |
| XM_017027064.1 | 2 | 287 | XP_016882553.1 | |
| AK097257.1 | 3 | 300 | ||
| BC111715.1 | 3 | 229 | AAI11716.1 | |
| BC111716.1 | 3 | 229 | AAI11717.1 | |
| D15049.2 | 3 | 247 | BAA03645.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv960874 | Chr.19:55202018 - 55210379 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map