Assay Details
Target Gene Details
Entrez Gene ID: | 196383 |
Gene Name: | Rab interacting lysosomal protein like 2 |
Gene Aliases: |
RLP2 |
Location: |
Chr.12:123408567-123436733 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RILPL2 | NM_145058.2 | 4 | 1083 | NP_659495.1 |
| NR_130703.1 | 3 | 931 | ||
| XM_011538012.2 | XP_011536314.1 | |||
| XM_017018959.1 | XP_016874448.1 | |||
| AB085763.1 | 4 | 846 | BAC76826.1 | |
| AK056934.1 | 4 | 1067 | BAB71313.1 | |
| BC013042.2 | 4 | 856 | AAH13042.1 | |
| BM556243.1 | 3 | 841 |
More Information
Set Membership: |
|

Genomic Map