Assay Details
Target Gene Details
Entrez Gene ID: | 64073 |
Gene Name: | chromosome 19 open reading frame 33 |
Gene Aliases: |
H2RSP, IMUP, IMUP-1, IMUP-2 |
Location: |
Chr.19:38304161-38305006 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C19orf33 | NM_001317801.1 | NP_001304730.1 | ||
| NM_033520.2 | NP_277055.1 | |||
| AB038317.1 | BAB18465.1 | |||
| AB038318.1 | BAB18466.1 | |||
| AF213678.1 | AAG43574.1 | |||
| BC014264.2 | ||||
| BC060319.1 | AAH60319.1 | |||
| BX648823.1 | ||||
| CB109881.1 |
Target Gene Details
Entrez Gene ID: | 90522 |
Gene Name: | Yip1 interacting factor homolog B, membrane trafficking protein |
Gene Aliases: |
FinGER8 |
Location: |
Chr.19:38303560-38315976 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIF1B | NM_001039671.2 | 8 | 1822 | NP_001034760.1 |
| NM_001039672.2 | 8 | 1840 | NP_001034761.1 | |
| NM_001039673.2 | 8 | 1831 | NP_001034762.1 | |
| NM_001145461.1 | 8 | 1816 | NP_001138933.1 | |
| NM_001145462.1 | 8 | 1792 | NP_001138934.1 | |
| NM_001145463.1 | 9 | 1908 | NP_001138935.1 | |
| AK027130.1 | 1 | 515 | BAB15669.1 | |
| BC025257.1 | 1 | 451 | AAH25257.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833822 | Chr.19:38270108 - 38454465 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map