Assay Details
Target Gene Details
Entrez Gene ID: | 146850 |
Gene Name: | phosphoinositide-3-kinase regulatory subunit 6 |
Gene Aliases: |
C17orf38, HsT41028, p84 PIKAP, p87(PIKAP), p87PIKAP |
Location: |
Chr.17:8802723-8870003 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3R6 | NM_001010855.3 | 11 | 1262 | NP_001010855.1 |
| NM_001290211.1 | 11 | 1630 | NP_001277140.1 | |
| NR_110865.1 | 10 | 1357 | ||
| XM_011523677.2 | 12 | 1275 | XP_011521979.1 | |
| XM_011523678.2 | 12 | 1259 | XP_011521980.1 | |
| XM_011523680.2 | 12 | 1275 | XP_011521982.1 | |
| XM_011523681.2 | 11 | 1523 | XP_011521983.1 | |
| XM_011523683.2 | 9 | 960 | XP_011521985.1 | |
| XM_011523686.2 | 3 | 409 | XP_011521988.1 | |
| XM_017024228.1 | 12 | 1274 | XP_016879717.1 | |
| XM_017024229.1 | 12 | 1274 | XP_016879718.1 | |
| XM_017024230.1 | 12 | 1274 | XP_016879719.1 | |
| XM_017024231.1 | 8 | 1451 | XP_016879720.1 | |
| AK091819.1 | 10 | 1359 | ||
| AY753192.1 | 9 | 1022 | AAV30085.1 | |
| BC125169.1 | 6 | 666 |
More Information
Set Membership: |
|

Genomic Map