Assay Details
Target Gene Details
Entrez Gene ID: | 164592 |
Gene Name: | coiled-coil domain containing 116 |
Gene Aliases: |
- |
Location: |
Chr.22:21632121-21637327 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC116 | NM_152612.2 | NP_689825.2 | ||
| XM_006724159.2 | XP_006724222.1 | |||
| XM_011529984.2 | XP_011528286.1 | |||
| XM_011529985.1 | XP_011528287.1 | |||
| AK093365.1 | BAC04145.1 | |||
| AK302030.1 | ||||
| BC033499.1 | AAH33499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160730 | Chr.22:21373849 - 21633730 on Build GRCh38 | Deletion |
|
| nsv516806 | Chr.22:21628603 - 21642781 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map