Assay Details
Target Gene Details
Entrez Gene ID: | 80099 |
Gene Name: | chromosome 7 open reading frame 69 |
Gene Aliases: |
- |
Location: |
Chr.7:47795291-47819847 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C7orf69 | NM_001302627.1 | NP_001289556.1 | ||
| NM_025031.2 | NP_079307.2 | |||
| AK024728.1 | BAB14977.1 | |||
| BC113679.1 | ||||
| BC113681.1 | ||||
| DA711092.1 |
Target Gene Details
Entrez Gene ID: | 168507 |
Gene Name: | polycystin 1 like 1, transient receptor potential channel interacting |
Gene Aliases: |
PRO19563 |
Location: |
Chr.7:47774603-47958622 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 49 - Exon 50 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKD1L1 | NM_138295.3 | NP_612152.1 | ||
| XM_017011798.1 | XP_016867287.1 | |||
| AB061683.1 | BAB85807.1 | |||
| AY358757.1 | AAQ89117.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3613069 | Chr.7:47708002 - 47898700 on Build GRCh38 | Gain |
|
| esv3613065 | Chr.7:47596195 - 48032523 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map