Assay Details
Target Gene Details
Entrez Gene ID: | 131965 |
Gene Name: | methyltransferase like 6 |
Gene Aliases: |
- |
Location: |
Chr.3:15381275-15427580 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL6 | NM_001301790.1 | 5 | 879 | NP_001288719.1 |
| NM_001301792.1 | 5 | 900 | NP_001288721.1 | |
| NM_152396.3 | 6 | 1014 | NP_689609.2 | |
| XM_005264867.3 | 6 | 1014 | XP_005264924.1 | |
| XM_006712970.3 | 6 | 890 | XP_006713033.1 | |
| XM_006712972.3 | XP_006713035.1 | |||
| XM_011533357.2 | 6 | 958 | XP_011531659.1 | |
| XM_017005718.1 | 6 | 890 | XP_016861207.1 | |
| XM_017005719.1 | 5 | 872 | XP_016861208.1 | |
| XM_017005722.1 | 5 | 872 | XP_016861211.1 | |
| XM_017005723.1 | XP_016861212.1 | |||
| AK027185.1 | 2 | 127 | ||
| AK293370.1 | 5 | 872 | ||
| BC022400.1 | AAH22400.1 | |||
| BI913767.1 | ||||
| CX872865.1 | 3 | 383 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834622 | Chr.3:15397807 - 15587083 on Build GRCh38 | Gain |
|
| nsv834621 | Chr.3:15324275 - 15509456 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map