Assay Details
Target Gene Details
Entrez Gene ID: | 391109 |
Gene Name: | olfactory receptor family 10 subfamily K member 1 |
Gene Aliases: |
OR1-6 |
Location: |
Chr.1:158465562-158466503 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10K1 | NM_001004473.1 | NP_001004473.1 | ||
| BC137323.1 | 1 | 104 | ||
| BC137324.1 | 1 | 104 |
More Information
Set Membership: |
|

Genomic Map