Assay Details
Target Gene Details
Entrez Gene ID: | 55735 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C11 |
Gene Aliases: |
dJ126A5.1 |
Location: |
Chr.1:6634168-6701906 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC11 | NM_018198.3 | 16 | 2294 | NP_060668.2 |
| XM_017001745.1 | 15 | 2147 | XP_016857234.1 | |
| AF052158.1 | 1 | 244 | ||
| AF306695.1 | 16 | 2208 | ||
| AK001599.1 | 16 | 2203 | BAA91780.1 | |
| AK025491.1 | 6 | 968 | ||
| AK093709.1 | 4 | 1449 | ||
| AK095386.1 | 7 | 2016 | ||
| AL109978.1 | CAB53376.1 | |||
| AL833841.1 | 16 | 2168 | CAD38701.1 | |
| AY336749.1 | AAR02411.1 | |||
| BC006086.1 | 16 | 2190 | AAH06086.1 | |
| BC008772.1 | 16 | 2195 | AAH08772.1 | |
| BC014145.2 | AAH14145.1 |
Target Gene Details
Entrez Gene ID: | 90326 |
Gene Name: | THAP domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.1:6624645-6635586 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THAP3 | NM_138350.3 | 5 | 1676 | NP_612359.2 |
| XM_017002761.1 | 5 | 1613 | XP_016858250.1 | |
| AF086370.1 | ||||
| AK096217.1 | 5 | 1634 | BAC04727.1 |
More Information
Set Membership: |
|

Genomic Map