Assay Details
Target Gene Details
Entrez Gene ID: | 84699 |
Gene Name: | cAMP responsive element binding protein 3 like 3 |
Gene Aliases: |
CREB-H, CREBH, HYST1481 |
Location: |
Chr.19:4153601-4173054 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB3L3 | NM_001271995.1 | NP_001258924.1 | ||
| NM_001271996.1 | NP_001258925.1 | |||
| NM_001271997.1 | NP_001258926.1 | |||
| NM_032607.2 | NP_115996.1 | |||
| AB050902.1 | BAB47242.1 | |||
| AB073612.1 | BAD38649.1 | |||
| AK172839.1 | BAD18804.1 | |||
| AK313099.1 | ||||
| BC101504.1 | AAI01505.1 | |||
| BC101508.1 | AAI01509.1 | |||
| BC143609.1 | ||||
| DA210258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833716 | Chr.19:4121259 - 4274854 on Build GRCh38 | Loss |
|
| esv3643488 | Chr.19:4137148 - 4158957 on Build GRCh38 | Gain |
|
| esv2717979 | Chr.19:4005406 - 4159344 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map