Assay Details
Target Gene Details
Entrez Gene ID: | 9703 |
Gene Name: | KIAA0100 |
Gene Aliases: |
BCOX, BCOX1, CT101, FMP27 |
Location: |
Chr.17:28614440-28645159 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA0100 | NM_014680.3 | 21 | 4054 | NP_055495.2 |
| XM_005258073.2 | 21 | 4049 | XP_005258130.1 | |
| XM_017025423.1 | 20 | 3903 | XP_016880912.1 | |
| BC120873.1 | 21 | 3913 | ||
| CR933634.1 | 22 | 3916 | CAI45940.1 | |
| D43947.3 | 21 | 4026 | BAA07891.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|

Genomic Map