Assay Details
Target Gene Details
Entrez Gene ID: | 2644 |
Gene Name: | GTP cyclohydrolase I feedback regulator |
Gene Aliases: |
GFRP, HsT16933, P35 |
Location: |
Chr.15:40764087-40767713 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCHFR | NM_005258.2 | NP_005249.1 | ||
| AK223430.1 | BAD97150.1 | |||
| AK311872.1 | ||||
| BC112262.1 | AAI12263.1 | |||
| BC112264.1 | AAI12265.1 | |||
| BC143904.1 |
More Information
Set Membership: |
|

Genomic Map