Assay Details
Target Gene Details
Entrez Gene ID: | 26493 |
Gene Name: | olfactory receptor family 8 subfamily B member 8 |
Gene Aliases: |
TPCR85 |
Location: |
Chr.11:124440150-124441085 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR8B8 | NM_012378.1 | 1 | 641 | NP_036510.1 |
| AF238488.1 | 1 | 641 | AAL32997.1 | |
| BC130393.1 | 1 | 714 | ||
| BC130419.1 | 1 | 714 | ||
| X89675.1 | 1 | 267 | CAA61822.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892105 | Chr.11:124309052 - 124530715 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map