Assay Details
Target Gene Details
Entrez Gene ID: | 79596 |
Gene Name: | ring finger protein 219 |
Gene Aliases: |
C13orf7 |
Location: |
Chr.13:78614286-78659179 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF219 | NM_024546.3 | NP_078822.3 | ||
| XM_011535225.1 | XP_011533527.1 | |||
| XM_017020743.1 | 2 | 200 | XP_016876232.1 | |
| AK098640.1 | ||||
| AL136651.1 | CAB66586.3 | |||
| BC136764.1 | ||||
| BC136765.1 | ||||
| BI550796.1 | ||||
| CR936752.1 | ||||
| CR936754.1 | ||||
| CR936764.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv339n67 | Chr.13:78560929 - 78651758 on Build GRCh38 | Gain |
|
| esv3580774 | Chr.13:78644908 - 78647311 on Build GRCh38 | Loss |
|
| nsv1098 | Chr.13:78622118 - 78655578 on Build GRCh38 | Insertion |
|
| nsv1142153 | Chr.13:78643555 - 78656350 on Build GRCh38 | Tandem Duplication |
|
More Information
Set Membership: |
|

Genomic Map