Assay Details
Target Gene Details
Entrez Gene ID: | 64168 |
Gene Name: | N-terminal EF-hand calcium binding protein 1 |
Gene Aliases: |
EFCBP1, STIP-1 |
Location: |
Chr.8:90791693-90959402 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECAB1 | NM_022351.4 | NP_071746.1 | ||
| XM_011517213.2 | XP_011515515.1 | |||
| AF414126.2 | AAP33146.1 | |||
| AK095533.1 | BAC04568.1 | |||
| AY224210.1 | AAO34126.1 | |||
| BC068448.1 | AAH68448.1 | |||
| DB481874.1 |
More Information
Set Membership: |
|

Genomic Map