Assay Details
Target Gene Details
Entrez Gene ID: | 26262 |
Gene Name: | tetraspanin 17 |
Gene Aliases: |
FBX23, FBXO23, TM4SF17 |
Location: |
Chr.5:176647387-176659058 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN17 | NM_001006616.2 | NP_001006617.2 | ||
| NM_012171.2 | NP_036303.1 | |||
| NM_130465.4 | NP_569732.2 | |||
| XM_005265876.3 | XP_005265933.1 | |||
| AF174603.1 | AAF04524.1 | |||
| AK024427.1 | BAB15717.1 | |||
| AY037146.1 | AAK67627.1 | |||
| BC010405.2 | AAH10405.1 | |||
| BC067105.1 | AAH67105.1 | |||
| BI756001.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600346 | Chr.5:176597879 - 176669235 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map